missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Novus Biologicals KSR1 Antibody (6C7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00008844-M01
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
KSR1 Monoclonal antibody specifically detects KSR1 in Human samples. It is validated for Western Blot, ELISA, ELISA
Tekniske data
| KSR1 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| kinase suppressor of ras, kinase suppressor of ras 1, KSRRSU2 | |
| KSR (XP_290793, 301 a.a. ~ 400 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. LDARRESGSGPSTDTLSAASLPWPPGSSQLGRAGNSAQGPRSISVSALPASDSPTPSFSEGLSDTCIPLHASGRLTPRALHSFITPPTTPQLRRHTKLKP | |
| 0.1 mg | |
| Protein Kinase, Signal Transduction | |
| 8844 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA, Sandwich ELISA | |
| 6C7 | |
| Western Blot, ELISA, Sandwich ELISA | |
| XP_290793 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion