Learn More
Novus Biologicals IQGAP1 Antibody (2C5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals H00008826-M01
Product Code. 18310699
  • 4545.00 DKK / 0.01mg
Explore more special offers
This item is not returnable. View return policy
Description
Description
IQGAP1 Monoclonal antibody specifically detects IQGAP1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), ELISA, KnockDown
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IQGAP1
Monoclonal
Unconjugated
In 1x PBS, pH 7.4
HUMORFA01, IQ motif containing GTPase activating protein 1, KIAA0051ras GTPase-activating-like protein IQGAP1, p195RasGAP-like with IQ motifs, SAR1
IQGAP1 (NP_003861, 611 a.a. ~ 710 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. WQSNKDTQEAQKFALGIFAINEAVESGDVGKTLSALRSPDVGLYGVIPECGETYHSDLAEAKKKKLAVGDNNSKWVKHWVKGGYYYYHNLETQEGGWDEP
0.1 mg
Signal Transduction
8826
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
IgG1 κ
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Sandwich ELISA, KnockDown
2C5
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Sandwich ELISA, Knockdown Validated
NP_003861
Mouse
IgG purified
RUO
Primary
Human
Purified