missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals Integrin alpha 9 Antibody (3E4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00003680-M01
Additional Details : Weight : 0.00970kg
Description
Integrin alpha 9 Monoclonal antibody specifically detects Integrin alpha 9 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
Integrin alpha 9 | |
Monoclonal | |
Unconjugated | |
In 1x PBS, pH 7.4 | |
ALPHA-RLC, integrin alpha-9, Integrin alpha-RLC, integrin, alpha 9, ITGA4L, RLC | |
ITGA9 (NP_002198, 785 a.a. ~ 886 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. GESVDAANFIQLDDLECHFQPINITLQVYNTGPSTLPGSSVSISFPNRLSSGGAEMFHVQEMVVGQEKGNCSFQKNPTPCIIPQEQENIFHTIFAFFTKSGR | |
0.1 mg | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
3E4 | |
Western Blot 1:500, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin | |
NP_002198 | |
Mouse | |
IgG purified | |
RUO | |
3680 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG1 κ |