missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals Hepsin Antibody (2D5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00003249-M02
Additional Details : Weight : 0.00970kg
Description
Hepsin Monoclonal antibody specifically detects Hepsin in Human samples. It is validated for Western Blot, ELISA, ELISASpecifications
Hepsin | |
Monoclonal | |
Unconjugated | |
In 1x PBS, pH 7.4 | |
EC 3.4.21, EC 3.4.21.106, hepsin, serine protease hepsin, serine protease hepsin catalytic chain, TMPRSS1, TMPRSS1serine protease hepsin non-catalytic chain, Transmembrane protease serine 1, transmembrane protease, serine 1 | |
HPN (AAH25716.1, 40 a.a. ~ 417 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. VAVLLRSDQEPLYPVQVSSADARLMVFDKTEGTWRLLCSSRSNARVAGLSCEEMGFLRALTHSELDVRTAGANGTSGFFCVDEGRLPHTQRLLEVISVCDCPRGRFLAAICQDCGRRKLPVDRIVGGRDTSLGRWPWQVSLRYDGAHLCGGSLLSGDWVLTAAHCFPERNRVLSRWRVFAGAVAQASPHGLQLGVQAVVYHGGYLPFRDPNSEENSNDIALVHLSSPLPLTEYIQPVCLPAAGQALVDGKICTVTGWGNTQYYGQQAGVLQEARVPIISNDVCNGADFYGNQIKPKMFCAGYPEGGIDACQGDSGGPFACEDSISRTPRWRLCGIVSWGTGCALAQKPGVYTKVSDFREWIFQAIKTHSEASGMVTQL | |
0.1 mg | |
Cancer, Prostate Cancer | |
3249 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG2a κ |
Western Blot, ELISA, Sandwich ELISA | |
2D5 | |
Western Blot 1:500, ELISA, Sandwich ELISA | |
AAH25716 | |
Mouse | |
IgG purified | |
RUO | |
Primary | |
Human | |
Purified |