missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals HECTD1 Antibody (1C7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00025831-M08
This item is not returnable.
View return policy
Description
HECTD1 Monoclonal antibody specifically detects HECTD1 in Human samples. It is validated for Western Blot, ELISA
Specifications
HECTD1 | |
Monoclonal | |
Unconjugated | |
In 1x PBS, pH 7.4 | |
E3 ligase for inhibin receptor, E3 ubiquitin-protein ligase HECTD1, EC 6.3.2.-, EULIR, HECT domain containing 1, HECT domain-containing protein 1, KIAA1131FLJ38315 | |
HECTD1 (NP_056197, 3 a.a. ~ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. DVDPDTLLEWLQMGQGDERDMQLIALEQLCMLLLMSDNVDRCFETCPPRTFLPALCKIFLDESAPDNVLEVTARAITYYLDVSAECTRRIVGVDGAIKALCNRLVVVE | |
0.1 mg | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
1C7 | |
Western Blot 1:500, ELISA | |
NP_056197 | |
Mouse | |
IgG purified | |
RUO | |
25831 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction