missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals Galactosylceramidase/GALC Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals H00002581-D01P
Additional Details : Weight : 0.00970kg
Description
Galactosylceramidase/GALC Polyclonal antibody specifically detects Galactosylceramidase/GALC in Human samples. It is validated for Western BlotSpecifications
Galactosylceramidase/GALC | |
Polyclonal | |
Western Blot | |
AAH36518.1 | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS (pH 7.4) | |
EC 3.2.1.46, galactocerebrosidase, Galactocerebroside beta-galactosidase, Galactosylceramidase, galactosylceramidase (Krabbe disease), Galactosylceramide beta-galactosidase, galactosylceraminidase, GALCERase | |
GALC (AAH36518.1, 1 a.a. - 669 a.a.) full-length human protein. MTAAAGSAGRAAVPLLLCALLAPGGAYVLDDSDGLGREFDGIGAVSGGGATSRLLVNYPEPYRSQILDYLFKPNFGASLHILKVEIGGDGQTTDGTEPSHMHYALDENYFRGYEWWLMKEAKKRNPNITLIGLPWSFPGWLGKGFDWPYVNLQLTAYYVVTWIVGAKRYHDLDIDYIGIWNERSYNANYIKILRKMLNYQGLQRVKIIASDNLWESISASMLLDAELFKVVDVIGAHYPGTHSAKDAKLTGKKLWSSEDFSTLNSDMGAGCWGRILNQNYINGYMTSTIAWNLVASYYEQLPYGRCGLMTAQEPWSGHYVVESPVWVSAHTTQFTQPGWYYLKTVGHLEKGGSYVALTDGLGNLTIIIETMSHKHSKCIRPFLPYFNVSQQFATFVLKGSFSEIPELQVWYTKLGKTSERFLFKQLDSLWLLDSDGSFTLSLHEDELFTLTTLTTGRKGSYPLPPKSQPFPSTYKDDFNVDYPFFSEAPNFADQTGVFEYFTNIEDPGEHHFTLRQVLNQRPITWAADASNTISIIGDYNWTNLTTKCDVYIETPDTGGVFIAGRVNKGGILIRSARGIFFWIFANGSYRVTGDLAGWIIYALGRVEVTAKKWYTLTLTIKGHFASGMLNDKSLWTDIPVNFPKNGWAAIGTHSFEFAQFDNFLVEATR | |
0.1 mg | |
Cancer, Cardiovascular Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
2581 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG |