missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Novus Biologicals FoxF1 Antibody (3D1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00002294-M05
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
FoxF1 Monoclonal antibody specifically detects FoxF1 in Human samples. It is validated for ELISA
Tekniske data
| FoxF1 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| FKHL5MGC105125, forkhead box F1, forkhead box protein F1, Forkhead, drosophila, homolog-like 5, Forkhead-related activator 1, Forkhead-related protein FKHL5, Forkhead-related transcription factor 1, FREAC-1, FREAC1ACDMPV | |
| FOXF1 (NP_001442, 251 a.a. ~ 353 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. AALNSGASYIKQQPLSPCNPAANPLSGSLSTHSLEQPYLHQNSHNAPAELQGIPRYHSQSPSMCDRKEFVFSFNAMASSSMHSAGGGSYYHQQVTYQDIKPCV | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 3D1 | |
| ELISA | |
| NP_001442 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 2294 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion