missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals FGF acidic/FGF1 Antibody (2E12), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00002246-M02
Additional Details : Weight : 0.00970kg
Description
FGF acidic/FGF1 Monoclonal antibody specifically detects FGF acidic/FGF1 in Human, Plant samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
FGF acidic/FGF1 | |
Monoclonal | |
Unconjugated | |
In 1x PBS, pH 7.4 | |
AFGF, alpha, ECGF, ECGFB, ECGF-betaAcidic fibroblast growth factor, endothelial cell growth factor, beta, FGFABeta-endothelial cell growth factor, FGF-alpha, fibroblast growth factor 1 (acidic), GLIO703, HBGF1, HBGF-1, heparin-binding growth factor 1 | |
FGF1 (AAH32697, 46 a.a. ~ 155 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. VDGTRDRSDQHIQLQLSAESVGEVYIKSTETGQYLAMDTDGLLYGSQTPNEECLFLERLEENHYNTYISKKHAEKNWFVGLKKNGSCKRGPRTHYGQKAILFLPLPVSSD | |
0.1 mg | |
Angiogenesis, Cancer | |
2246 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
2E12 | |
Western Blot 1:500, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin | |
AAH32697 | |
Mouse | |
IgG purified | |
RUO | |
Primary | |
Human, Plant | |
Purified |