missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Novus Biologicals BRD3 Antibody (6E7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00008019-M01
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
BRD3 Monoclonal antibody specifically detects BRD3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Tekniske data
| BRD3 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| bromodomain containing 3, bromodomain-containing protein 3, KIAA0043FLJ41328, ORFXbromodomain-containing 3, RING3LFLJ23227, RING3-like protein | |
| BRD3 (AAH32124, 418 a.a. ~ 556 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. EPVEAPALPAPAAPMVSKGAESSRSSEESSSDSGSSDSEEERATRLAELQEQLKAVHEQLAALSQAPVNKPKKKKEKKEKEKKKKDKEKEKEKHKVKAEEEKKAKVAPPAKQAQQKKAPAKKANSTTTAGRDHFLTCGV | |
| 0.1 mg | |
| Protein Kinase | |
| 8019 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Sandwich ELISA | |
| 6E7 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA | |
| AAH32124 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion