missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals ANKRD15 Antibody (2B8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00023189-M01
Additional Details : Weight : 0.00970kg
Description
ANKRD15 Monoclonal antibody specifically detects ANKRD15 in Human samples. It is validated for Western Blot, ELISA, ELISASpecifications
ANKRD15 | |
Monoclonal | |
Unconjugated | |
In 1x PBS, pH 7.4 | |
ANKRD15ankyrin repeat domain 15, Ankyrin repeat domain-containing protein 15, KANKDKFZp451G231, KIAA0172MGC43128, Kidney ankyrin repeat-containing protein, KN motif and ankyrin repeat domain-containing protein 1, KN motif and ankyrin repeat domains 1 | |
ANKRD15 (AAH38116, 701 a.a. ~ 800 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MGSLNSQLISTLSSINSVMKSASTEELRNPDFQKTSLGKITGNYLGYTCKCGGLQSGSPLSSQTSQPEQEVGTSEGKPISSLDAFPTQEGTLSPVNLTDD | |
0.1 mg | |
Cell Cycle and Replication | |
23189 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG2a κ |
Western Blot, ELISA, Sandwich ELISA | |
2B8 | |
Western Blot 1:500, ELISA, Sandwich ELISA | |
AAH38116 | |
Mouse | |
IgG purified | |
RUO | |
Primary | |
Human | |
Purified |