Honeywell Fluka Buffer solution pH 4.0 (20°C), Red colored, citric acid / sodium hydroxide / sodium chloride solution, with fungicide, Honeywell Fluka™
Red colored, citric acid / sodium hydroxide / sodium chloride solution, with fungicide
Additional Details:
Additional Details: Weight: 5.97000kg
Quantity 6 x 1 L
Product Code. 15674060
Quantity Price
1 3223.06  DKK / Pack of 6
In stock, ready to ship: 125
Add to basket
  • Description and Specification
  • Alternative Products
Description and Specification
Chemical Identifiers
CAS
Molecular Formula
Molecular Weight (g/mol)
MDL Number
InChI Key
PubChem CID
ChEBI
SMILES
Specification
Chemical Name or Material Buffer solution pH 4.0 (20°C)
Name Note Red colored, citric acid / sodium hydroxide / sodium chloride solution, with fungicide
Boiling Point 100°C
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
CAS 18996-35-5
CAS Max % <1.0000%
Density 1.0 g/cm3
CAS 144-33-2
CAS 7647-14-5
Quantity 6 x 1 L
Solution Type Buffer
Packaging Poly Bottle
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Alternative Products
Alternative Packaging
Product Code Brand Description Stock Price
Quantity
15684060 Honeywell Fluka X6 Buffer solution pH 4.0 (20oC) red colored, 500ML 38 1686.92 DKK
Add to basket
15664060 Honeywell Fluka 5LT Buffer solution pH 4.0 (20oC) red colored, citric acid / sodium hydroxide / sodium chloride Estimated Shipment
03-09-2025
921.73 DKK
Add to basket
15634060 Honeywell Fluka 10LT Buffer solution pH 4.0 (20oC) red colored, citric acid / sodium hydroxide / sodium chloride Estimated Shipment
31-07-2025
2256.19 DKK
Add to basket
Alternative Technical
Product Code Brand Description Stock Price
Quantity
15644060 Honeywell Fluka 1LT Buffer solution pH 4.0 (20oC) red colored, 751 618.22 DKK
Add to basket
Other Alternatives
Product Code Brand Description Stock Price
Quantity
15624030 Honeywell Fluka X6 Buffer solution pH 4.0 (20oC) with fungicide,1L 158 2451.97 DKK
Add to basket