UserName
Every Space On the Bench
Equip your lab bench with the right tools and enhance your work.
The Perfect Products to Complement Your Lab Bench
Our comprehensive range of on the bench essentials is specifically designed to cater to your needs, ensuring efficiency and flexibility in every aspect of your work.
Green Solutions
Energy Efficiency
Highlighting 6 ways to help make your lab more sustainable.
FAQ
When selecting a benchtop centrifuge, consider factors such as capacity, speed range, rotor compatibility, and safety features. Assess your lab's specific needs to find the best fit.
Microcentrifuges are specialized centrifuges designed for small volume samples, typically ranging from microliters to milliliters. They operate at high speeds to generate centrifugal force for applications such as DNA/RNA isolation, protein precipitation, and cell pelleting, making them ideal for molecular biology and biochemistry research.
Benchtop pH meters are stationary instruments designed for precise and accurate measurement of pH in laboratory settings. They typically offer advanced features such as temperature compensation, data logging, and compatibility with specialized electrodes, making them suitable for demanding applications requiring high accuracy and reproducibility.
Each parameter can be toggled on/off independently.
Lab Reporter
Other Programs
Sustainability Program
Read more about how we can reach net-zero goals together.
Edge banner
Fisher Scientific Edge Program
Ensure quick dispatch of our most popular products.
Edge banner
Setting Up a New Lab?
Get all the support needed for setting up a new lab or expanding an existing lab through the NLSU program.
Edge banner
News Corner
Explore the latest innovations within the Fisher Scientific Channel.
Edge banner