UserName
Sustainability Recycling Programs
Sustainability Recycling Program
Recycling Programs for a Cleaner Tomorrow
From programs for recycling pipette tips to garment recycling, we have solutions developed with your sustainability goals in mind.
DuPont Tyvek Garment Recycling
The DuPont™ Tyvek™ protective apparel recycling program diverts garments away from landfill and gives them a second life as new products like containers, pallets, and park benches.
Availability
Austria, Belgium, Denmark, Germany, Netherlands, UK
DuPont Tyvek Garment Recycling
RightCycle Program
The RightCycle™ Program is a large-scale recycling program, turning non-hazardous nitrile and latex gloves, safety spectacles, and apparel into new plastic products and durable goods.
Availability
Austria, Belgium, France, Germany, Ireland, Spain, Netherlands, Switzerland, UK
RightCycle Program
ReNew by Kimberly-Clark Professional™
Instead of sending used hand towels, old dispensers, and wipers to landfill or incineration, the ReNew program collects them from your business and recycles them into new consumer goods. Learn More
Availability
  • Hand Towels: Germany, UK 
  • Dispensers: Germany, UK, Netherlands, France 
  • Wipers: Germany, UK coming soon
ReNew by Kimberly-Clark ProfessionalTM
Fisher Scientific Recycling Solutions
Find Recycling Programs and Responsible End-of-Use Options for Your Products
Circularity is a transformative approach to sustainability, where products are designed, used, and recycled continuously, minimizing waste and conserving resources. Our guide helps you manage the end-of-use phase of your products by providing information on various plastics and materials, along with information on where and who can recycle them. We make recycling accessible by helping you locate programs and major recyclers in your area.
Our aim is to provide you with the knowledge and resources needed to help support getting your products recycled and repurposed effectively. Our take-back programs reclaim instruments and components, avoiding wasteful disposal and meeting global recycling requirements. Let us help you recycle, preventing excess waste from occupying space in your lab or landfill.
Our guidance covers the recycling of plastic labware and instruments deemed non-hazardous according to local and country-specific regulations. The listed recyclers have a broad regional presence, helping to ensure accessibility for many users. Detailed instructions and further information on recycling can be obtained from the respective recycling programs and providers. This guidance currently applies to the United States and selected European countries only.*
Together, we can close the loop and build a more sustainable future.
Key to Acronyms, Resin Identification Codes, and Common Names for Plastics Types
ABS (#7): Acrylonitrile butadiene styrene
EPS (#6): Expanded polystyrene
FEP (#7): Fluorinated ethylene propylene 
HDPE (#2): High-density polyethylene
LDPE (#4): Low-density polyethylene
PC (#7): Polycarbonate
PE: Polyethylene 
PES: Polyether sulfone
PET/PETE (#1): Polyethylene terephthalate 
PETG (#7): Polyethylene terephthalate glycol
PFA (#7): Perfluoroalkoxy
PTFE (#7): Polytetrafluoroethylene
PP (#5): Polypropylene
PPCO (#5): Polypropylene copolymer 
PS (#6): Polystyrene
EPE:  Expanded polyethylene
EPU:  Expanded polyurethane
TPE: Thermoplastic elastomers 
Key to Acronyms
Labware and Supplies
Labware and Supplies
Lab Equipment, Instruments, and Accessories
Lab Equipment, Instruments, and Accessories
Safety
Safety Products
Packaging
Packaging
Compliance with the European Union’s Waste Electrical & Electronic Equipment (WEEE) Directive
Compliance with the European Union’s Waste Electrical and Electronic Equipment (WEEE) Directive
Products with this symbol are required to comply with the European Union’s Waste Electrical and Electronic Equipment (WEEE) Directive 2012/19/EU.
Contact your sales representative to learn more about Fisher Scientific equipment and instrument trade-in, return, and recycling programs.
*Thermo Fisher Scientific and its affiliates make no assertions or guarantees as to the services/products that third-party recyclers offer or their accuracy with regard to processes, results, or outcomes. Thermo Fisher Scientific takes no legal or financial responsibility for services, products, or the statements made by third-party recyclers, either on this page or in its marketing materials. Each customer of these recyclers must make its own determinations in whether to utilize these entities for their intended purposes.
For Research Use Only. Not for use in diagnostic procedures.