CAS RN 9045-22-1
CAS RN 9045-22-1
IUPAC Name:
3-phenylprop-2-enoic acid
Synonyms:
phenylacrylic acid zimtsaeure
Molecular Weight (g/mol):
148.16
Molecular Formula:
C9H8O2
InChi Key:
WBYWAXJHAXSJNI-UHFFFAOYSA-N
SMILES:
OC(=O)C=CC1=CC=CC=C1
1
–
1
of
1
results
1
Medchem Express Heparin Lithium salt, MedChemExpress
MedChemExpress Heparin Lithium salt is an anticoagulant which binds reversibly to antithrombin III (ATIII). Heparin Lithium salt significantly inhibits exosome-cell interactions.
