accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 636-00-0

HBrNH2OHOHHO

CAS RN 636-00-0

IUPAC Name: hydrogen 5-(2-aminoethyl)benzene-1,2,4-triol bromide
Synonyms: hydrogen hydroxydopamine bromide
Molecular Weight (g/mol): 250.09
Molecular Formula: C8H12BrNO3
InChi Key: MLACDGUOKDOLGC-UHFFFAOYSA-N
SMILES: [H+].[Br-].NCCC1=CC(O)=C(O)C=C1O
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Grade 
  • (5)

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (5)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (5)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.