CAS RN 62-75-9
CAS RN 62-75-9
IUPAC Name:
dimethyl(nitroso)amine
Synonyms:
nitrosodimethylamine DMNA ndma nitrosodimethylamine rcra waste number P082 DMN nitrosodimethylamine, ndma ccris 261 N-nitroaodimethylamine
Molecular Weight (g/mol):
74.08
Molecular Formula:
C2H6N2O
InChi Key:
UMFJAHHVKNCGLG-UHFFFAOYSA-N
SMILES:
CN(C)N=O