CAS RN 58579-51-4
CAS RN 58579-51-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Anagrelide Hydrochloride, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Anagrelide hydrochloride, MedChemExpress
MedChemExpress Anagrelide hydrochloride (BL4162A) is a drug used for the treatment of essential thrombocytosis.
