accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 542-91-6

CH3H2SiH3C

CAS RN 542-91-6

IUPAC Name: diethylsilane
Synonyms: silane, diethyl-
Molecular Weight (g/mol): 88.23
Molecular Formula: C4H12Si
InChi Key: UCXUKTLCVSGCNR-UHFFFAOYSA-N
SMILES: CC[SiH2]CC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.