accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 528-29-0

ONONOO

CAS RN 528-29-0

IUPAC Name: 1,2-dinitrobenzene
Synonyms: O-dinitrobenzene
Molecular Weight (g/mol): 168.11
Molecular Formula: C6H4N2O4
InChi Key: IZUKQUVSCNEFMJ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1[N+]([O-])=O
Concentration 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.