accessibility menu, dialog, popup

UserName

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 518-28-5

CH3OOCH3OH3C(R)(R)(R)OO(R)OHOO

CAS RN 518-28-5

IUPAC Name: (10R,11R,15R,16R)-16-hydroxy-10-(3,4,5-trimethoxyphenyl)-4,6,13-trioxatetracyclo[7.7.0.0³,⁷.0¹¹,¹⁵]hexadeca-1,3(7),8-trien-12-one
Synonyms: podofilox podophyllotoxin 7 condylox podophyllinic acid lactone podophyllotoxin
Molecular Weight (g/mol): 414.41
Molecular Formula: C22H22O8
InChi Key: YJGVMLPVUAXIQN-XVVDYKMHSA-N
SMILES: COC1=CC(=CC(OC)=C1OC)[C@H]1[C@@H]2[C@H](COC2=O)[C@@H](O)C2=CC3=C(OCO3)C=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.