CAS RN 500-64-1
CAS RN 500-64-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
(+)-Kavain, MedChemExpress
MedChemExpress (+)-Kavain, a main kavalactone extracted from Piper methysticum, has anticonvulsive properties, attenuating vascular smooth muscle contraction through interactions with voltage-dependent Na+ and Ca2+ channels. (+)-Kavain is shown to bind at the α4β2δ GABAA receptor and potentiate GABA efficacy. (+)-Kavain is used as a treatment for inflammatory diseases, its anti-inflammatory action has been widely studied.