accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 40596-69-8

CH3OCH3CH3CH3CH3OOCH3H3C(E)(E)

CAS RN 40596-69-8

IUPAC Name: propan-2-yl (2E,4E)-11-methoxy-3,7,11-trimethyldodeca-2,4-dienoate
Synonyms: manta precor caswell no. 028aaa apex (pesticide) 甲氧普烯 methoprene S altosid PS 10 juvemon minex kabat
Molecular Weight (g/mol): 310.48
Molecular Formula: C19H34O3
InChi Key: NFGXHKASABOEEW-LDRANXPENA-N
SMILES: COC(C)(C)CCCC(C)C\C=C\C(\C)=C\C(=O)OC(C)C
Molecular Weight (g/mol) 
  • (2)
  • (1)
  • (1)
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.