CAS RN 37147-15-2
CAS RN 37147-15-2
IUPAC Name:
5-(ethylsulfanyl)-2,3-dihydro-1,3,4-thiadiazole-2-thione
Synonyms:
5-(ethylsulfanyl)-3H-1,3,4-thiadiazole-2-thione
Molecular Weight (g/mol):
178.29
Molecular Formula:
C4H6N2S3
InChi Key:
JLJYQXMUUKOBBO-UHFFFAOYSA-N
SMILES:
CCSC1=NNC(=S)S1
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar 2-Ethylthio-1,3,4-thiadiazole-5-thiol, 97%, Thermo Scientific Chemicals
CAS: 37147-15-2 Molecular Formula: C4H6N2S3 Molecular Weight (g/mol): 178.29 MDL Number: MFCD00186131 InChI Key: JLJYQXMUUKOBBO-UHFFFAOYSA-N Synonym: 2-ethylthio-1,3,4-thiadiazole-5-thiol,5-ethylthio-1,3,4-thiadiazole-2-thiol,5-ethylsulfanyl-1,3,4-thiadiazole-2-thiol,2-ethylthio-1,3,4-thiazole-5-thiol,5-ethylthio-1,3,4-thiadiazole-2 3h-thione,5-ethylsulfanyl-1,3,4-thiadiazole-2 3h-thione,acmc-1cszq,5-ethylthio-3h-1,3,4-thiadiazole-2-thione PubChem CID: 1201366 IUPAC Name: 5-ethylsulfanyl-3H-1,3,4-thiadiazole-2-thione SMILES: CCSC1=NNC(=S)S1