CAS RN 370586-05-3
CAS RN 370586-05-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express CID-797718, MedChemExpress
MedChemExpress CID-797718 is a compound with unknown details.
