accessibility menu, dialog, popup

UserName

Save up to 60% on cold storage essentials

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 3068-32-4

CH3OOOBrOCH3OOCH3OOCH3O

CAS RN 3068-32-4

IUPAC Name: [3,4,5-tris(acetyloxy)-6-bromooxan-2-yl]methyl acetate
Synonyms: [3,4,5-tris(acetyloxy)-6-bromooxan-2-yl]methyl acetate
Molecular Weight (g/mol): 411.2
Molecular Formula: C14H19BrO9
InChi Key: CYAYKKUWALRRPA-UHFFFAOYNA-N
SMILES: CC(=O)OCC1OC(Br)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.