CAS RN 2782024-58-0
CAS RN 2782024-58-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscienceâ„¢ Phenyl-glutarimide 4'-oxyacetic acid
Functionalized cereblon ligand for PROTAC development