CAS RN 24880-40-8
CAS RN 24880-40-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
(all-cis)-8,11,14,17-Eicosatetraenoic Acid (10 mg/mL ethanol), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.