CAS RN 2484876-94-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
R&D Systems™ NAMPT PROTAC™ A7
A Tocris product, part of R&D Systems. Potent and selective nicotinamide phosphoribosyl transferase (NAMPT) Degrader (PROTAC™;)