CAS RN 2483-57-0
IUPAC Navn:
methyl 2-nitroacetate
Synonymer:
nitroacetic acid methyl ester
Molekylvægt (g/mol):
119.08
Molekylformel:
C3H5NO4
InChi Key:
ALBSWLMUHHZLLR-UHFFFAOYSA-N
SMILES:
COC(=O)C[N+]([O-])=O
1
–
1
af
1
Resultater
1
Methylnitroacetat, 97+%
CAS: 2483-57-0 Molekylær formel: C3H5NO4 Molekylvægt (g/mol): 119.076 MDL nummer: MFCD00075480 InChI nøgle: ALBSWLMUHHZLLR-UHFFFAOYSA-N Synonym: methyl nitroacetate,nitroacetic acid methyl ester,acetic acid, nitro-, methyl ester,methylnitroacetate,methyl nitrozcetate,methyl nitro acetate,zlchem 149,methyl a-nitroacetate,methyl 2-nitroethanoate,acmc-1clk8 PubChem CID: 17206 IUPAC navn: methyl-2-nitroacetat SMIL: COC(=O)C[N+](=O)[O-]