CAS RN 247940-06-3
CAS RN 247940-06-3
IUPAC Navn:
{[1,1'-biphenyl]-2-yl}dicyclohexylphosphane
Synonymer:
[1,1'-biphenyl]-2-yldicyclohexylphosphane
Molekylvægt (g/mol):
350.49
Molekylformel:
C24H31P
InChi Key:
LCSNDSFWVKMJCT-UHFFFAOYSA-N
SMILES:
C1CCC(CC1)P(C1CCCCC1)C1=CC=CC=C1C1=CC=CC=C1
1
–
3
af
3
Resultater
1
2-(Dicyclohexylphosphino)biphenyl, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-(Dicyclohexylphosphino)biphenyl, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-(Dicyclohexylphosphino)biphenyl, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.