CAS RN 2470-73-7
CAS RN 2470-73-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Dixyrazine, MedChemExpress
MedChemExpress Dixyrazine, a phenothiazine derivative, can prevent brain oedema induced by intracarotid injection of protamine sulphate.