CAS RN 2468656-63-3
CAS RN 2468656-63-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
7-(But-2-yn-1-yl)-3-methyl-8-(methylamino)-1-((4-methylquinazolin-2-yl)methyl)-1H-purine-2,6(3H,7H)-dione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
7-(But-2-yn-1-yl)-3-methyl-8-(methylamino)-1-((4-methylquinazolin-2-yl)methyl)-1H-purine-2,6(3H,7H)-dione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.