CAS RN 2467-02-9
IUPAC Navn:
2-[(2-hydroxyphenyl)methyl]phenol
Synonymer:
bisphenol F 2,2'-bisphenol F
Molekylvægt (g/mol):
200.24
Molekylformel:
C13H12O2
InChi Key:
MQCPOLNSJCWPGT-UHFFFAOYSA-N
SMILES:
OC1=CC=CC=C1CC1=CC=CC=C1O
1
–
2
af
2
Resultater
1
2,2'-Bis(hydroxyphenyl)methane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,2'-Bis(hydroxyphenyl)methane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.