CAS RN 2459446-33-2
CAS RN 2459446-33-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Benzyl (2S)-3-Methyl-2-[pentanoyl[[2'-[2-(triphenylmethyl)tetrazol-5-yl]biphenyl-4-yl]methyl]amino]butanoate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.