CAS RN 24587-49-3
CAS RN 24587-49-3
IUPAC Navn:
4-hydroxybut-2-enoic acid
Synonymer:
4-hydroxy-2-butenoic acid
Molekylvægt (g/mol):
102.09
Molekylformel:
C4H6O3
InChi Key:
RMQJECWPWQIIPW-UHFFFAOYSA-N
SMILES:
OCC=CC(O)=O
1
–
1
af
1
Resultater
1
(E)-4-Hydroxycrotonic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.