accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 18087-73-5

NNNBr

CAS RN 18087-73-5

IUPAC Name: 3-bromoimidazo[1,2-b]pyridazine
Synonyms: 3-bromoimidazo[1,2-b]pyridazine
Molecular Weight (g/mol): 198.02
Molecular Formula: C6H4BrN3
InChi Key: KJQVHOFAWISYDO-UHFFFAOYSA-N
SMILES: BrC1=CN=C2C=CC=NN12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.