accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 173341-02-1

CH3H3CCH3OOHNNHO

CAS RN 173341-02-1

IUPAC Name: tert-butyl N-[(morpholin-2-yl)methyl]carbamate
Synonyms: tert-butyl N-(morpholin-2-ylmethyl)carbamate
Molecular Weight (g/mol): 216.28
Molecular Formula: C10H20N2O3
InChi Key: IYHJNCQAADULQE-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NCC1CNCCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.