accessibility menu, dialog, popup

UserName

The Ultimate Lab Appliances Promotion

Save Up to 78%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 16063-80-2

NH2FFFFFFOHO

CAS RN 16063-80-2

IUPAC Name: 2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butanoic acid
Synonyms: 2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butanoic acid
Molecular Weight (g/mol): 225.09
Molecular Formula: C5H5F6NO2
InChi Key: KRNSHCKTGFAMPQ-UHFFFAOYNA-N
SMILES: NC(C(C(F)(F)F)C(F)(F)F)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.