accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 14611-34-8

H2NNOONHNOOHOOOO(E/Z)

CAS RN 14611-34-8

IUPAC Name: 5-{[amino({[(benzyloxy)carbonyl]imino})methyl][(benzyloxy)carbonyl]amino}-2-{[(benzyloxy)carbonyl]amino}pentanoic acid
Synonyms: 5-{[amino({[(benzyloxy)carbonyl]imino})methyl][(benzyloxy)carbonyl]amino}-2-{[(benzyloxy)carbonyl]amino}pentanoic acid
Molecular Weight (g/mol): 576.61
Molecular Formula: C30H32N4O8
InChi Key: YSGAXJCIEJGVFV-UHFFFAOYNA-N
SMILES: NC(=NC(=O)OCC1=CC=CC=C1)N(CCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O)C(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.