CAS RN 1362850-20-1
CAS RN 1362850-20-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express Seletalisib, MedChemExpress
MedChemExpress Seletalisib (UCB5857) is potent and selective PI3Kδ inhibitor with an IC50 of 12 nM.

