CAS RN 123334-23-6
CAS RN 123334-23-6
IUPAC Name:
trisodium hexachloroiridiumtriuide hydrate
Synonyms:
trisodium hexachloroiridiumtriuide hydrate
Molecular Weight (g/mol):
491.9
Molecular Formula:
Cl6H2IrNa3O
InChi Key:
DXIIVDVTQZORNM-UHFFFAOYSA-H
SMILES:
O.[Na+].[Na+].[Na+].Cl[Ir-3](Cl)(Cl)(Cl)(Cl)Cl
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar Sodium hexachloroiridate(III) hydrate, Ir 31.5% min
CAS: 123334-23-6 Molecular Formula: Cl6IrNa3 Molecular Weight (g/mol): 473.89 MDL Number: MFCD00003508 InChI Key: GSONQQIOULBMRS-UHFFFAOYSA-H Synonym: sodium hexachloroiridate iii hydrate,sodium hexachloroiridate 3--water 3/1/1,cl6ir.3na.h2o,sodium hexachloroiridate iii hydrate, ir,iridate 3-,hexachloro-, sodium, hydrate 1:3:? , oc-6-11 PubChem CID: 56845692 IUPAC Name: trisodium;hexachloroiridium(3-);hydrate SMILES: [Na+].[Na+].[Na+].Cl[Ir-3](Cl)(Cl)(Cl)(Cl)Cl