accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 121-82-4

CAS RN 121-82-4

IUPAC Name: 1,3,5-trinitro-1,3,5-triazinane
Synonyms: cyclotrimethylenetrinitramine hexogen RDX cyclonite
Molecular Weight (g/mol): 222.12
Molecular Formula: C3H6N6O6
InChi Key: XTFIVUDBNACUBN-UHFFFAOYSA-N
SMILES: [O-][N+](=O)N1CN(CN(C1)[N+]([O-])=O)[N+]([O-])=O
Quantity 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.