accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 118430-74-3

H3CNNNH2

CAS RN 118430-74-3

IUPAC Name: 3-cyclopropyl-1-methyl-1H-pyrazol-5-amine
Synonyms: 5-cyclopropyl-2-methylpyrazol-3-amine
Molecular Weight (g/mol): 137.19
Molecular Formula: C7H11N3
InChi Key: YWHWPIRLFHZSFS-UHFFFAOYSA-N
SMILES: CN1N=C(C=C1N)C1CC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.