CAS RN 112883-39-3
CAS RN 112883-39-3
IUPAC Name:
4-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-oxobutanoic acid
Synonyms:
4-(tert-butoxy)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-oxobutanoic acid
Molecular Weight (g/mol):
411.45
Molecular Formula:
C23H25NO6
InChi Key:
FODJWPHPWBKDON-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar N-Fmoc-D-aspartic acid 4-tert-butyl ester, 98%
CAS: 112883-39-3 Molecular Formula: C23H25NO6 Molecular Weight (g/mol): 411.45 MDL Number: MFCD00077050 InChI Key: FODJWPHPWBKDON-UHFFFAOYNA-N Synonym: fmoc-d-asp otbu-oh,r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy-4-oxobutanoic acid,fmoc-d-asp obut-oh,fmoc-d-aspartic acid beta-tert-butyl ester,n-alpha-fmoc-d-aspartic acid beta-t-butyl ester,2r-4-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid,fmoc-dasp otbu,ambotzfaa1309,fmoc-dasp otbu-oh,pubchem10009 PubChem CID: 7018816 SMILES: CC(C)(C)OC(=O)CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O