CAS RN 112362-50-2
CAS RN 112362-50-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Dalfopristin, MedChemExpress
MedChemExpress Dalfopristin is a semi-synthetic streptogramin antibiotic. Quinupristin/Dalfopristin (Q/D) is a valuable alternative antibiotic to vancomycin for the treatment of multi-drug resistant Enterococcus faecium infections.