accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1083-30-3

O

CAS RN 1083-30-3

IUPAC Name: 1,3-diphenylpropan-1-one
Synonyms: dihydrochalcone
Molecular Weight (g/mol): 210.28
Molecular Formula: C15H14O
InChi Key: QGGZBXOADPVUPN-UHFFFAOYSA-N
SMILES: O=C(CCC1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.