CAS RN 107753-78-6
CAS RN 107753-78-6
IUPAC Navn:
cyclopentyl N-[3-({2-methoxy-4-[(2-methylbenzenesulfonyl)carbamoyl]phenyl}methyl)-1-methyl-1H-indol-5-yl]carbamate
Synonymer:
zafirlukast aeronix menarini brand of zafirlukast zafirlukast (product) novartis brand of zafirlukast accolate astrazeneca brand of zafirlukast zafirlukast (substance) zafirlukast preparation zeneca brand of zafirkulast
Molekylvægt (g/mol):
575.68
Molekylformel:
C31H33N3O6S
InChi Key:
YEEZWCHGZNKEEK-UHFFFAOYSA-N
SMILES:
COC1=CC(=CC=C1CC1=CN(C)C2=CC=C(NC(=O)OC3CCCC3)C=C12)C(=O)NS(=O)(=O)C1=CC=CC=C1C
1
–
2
af
2
Resultater
1
Zafirlukast, MedChemExpress
MedChemExpress Zafirlukast (ICI 204219) is a potent orally active leukotriene D4 (LTD4) receptor antagonist. Zafirlukast shows anti-asthmatic, anti-inflammatory and anti-bacterial effects.