accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1052532-15-6

H3CONHONClOCH3OCH3HCl

CAS RN 1052532-15-6

IUPAC Name: ({4-[(6-chloropyridin-3-yl)methoxy]-3-methoxyphenyl}methyl)[2-(3,4-dimethoxyphenyl)ethyl]amine hydrochloride
Synonyms: ({4-[(6-chloropyridin-3-yl)methoxy]-3-methoxyphenyl}methyl)[2-(3,4-dimethoxyphenyl)ethyl]amine hydrochloride
Molecular Weight (g/mol): 479.4
Molecular Formula: C24H28Cl2N2O4
InChi Key: QBGSVDJLQQXEGG-UHFFFAOYSA-N
SMILES: Cl.COC1=C(OC)C=C(CCNCC2=CC(OC)=C(OCC3=CN=C(Cl)C=C3)C=C2)C=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.