CAS RN 104757-53-1
CAS RN 104757-53-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Barnidipine hydrochloride, MedChemExpress
MedChemExpress Barnidipine hydrochloride (Mepirodipine hydrochloride) is an L-type calcium antagonist (CaA) with high affinity for [3H] initrendipine binding sites (Ki=0.21 nmol/l), has selective action against CaA receptors. Barnidipine hydrochloride (Mepirodipine hydrochloride) is an antihypertensive drug and acts by the reduction of peripheral vascular resistance secondary to its vasodilatory action.
