CAS RN 104632-27-1
CAS RN 104632-27-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dexpramipexole dihydrochloride, MedChemExpress
MedChemExpress Dexpramipexole dihydrochloride ((R)-Pramipexole dihydrochloride) is a neuroprotective agent and weak non-ergoline dopamine agonist.
